missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ethenesulfonate (25% in H2O), TRC

Product Code. 30416313
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
Unit Size:
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416313 10 mL -
30416709 5 mL -
30410373 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30416313 Supplier Toronto Research Chemicals Supplier No. TRCS69939810ML
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3039-83-6
Molecular Formula C2H3NaO3S
Molecular Weight (g/mol) 130.1
Synonym HS 1, HS 1 (Salt), N-SVS 25, Provichem 2202, SVS, SVS (Emulsifier), SVS 25, Sodium Ethenesulfonate, Sodium Ethylenesulfonate, Sodium Vinyl Sulphonate, Sodium Vinylsulfonate, Tamol VS, Vinalyst 3525, Vinylsulfonic Acid Sodium Salt
IUPAC Name sodium;ethenesulfonate
SMILES [Na+].[O-]S(=O)(=O)C=C

Specifications

Quantity 10 mL
InChI Formula InChI=1S/C2H4O3S.Na/c1-2-6(3,4)5;/h2H,1H2,(H,3,4,5);/q;+1/p-1
Formula Weight 129.9701
Concentration or Composition (by Analyte or Components) 25%
Chemical Name or Material Sodium Ethenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.