missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™

Catalog Number 10356463
164.96 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


171.12 GBP valid until 2024-12-31
Use promo code "23286" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Glass Bottle
Poly Bottle
Poly Pail
C12H25NaO4S
288.38
MFCD00036175
DBMJMQXJHONAFJ-UHFFFAOYSA-M
Sodium Lauryl Sulfate, SDS
3423265
CHEBI:8984
sodium dodecyl sulfate
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
C12H25NaO4S
288.38
MFCD00036175
DBMJMQXJHONAFJ-UHFFFAOYSA-M
Sodium Lauryl Sulfate, SDS
3423265
CHEBI:8984
sodium dodecyl sulfate
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

For protein extraction procedures and SDS polyacrylamide gels

  • DNase and RNase-free detergent for protein extraction procedures and polyacrylamide gel electrophoresis
  • SDS is a commonly used detergent in protein purification and polyacrylamide gel electrophoresis (PAGE)
  • SDS denatures proteins by disrupting the non-covalent bonds of the protein
  • Functionally tested for use in electrophoresis
  • Intended for research use only; not for human or animal diagnostic purposes
TRUSTED_SUSTAINABILITY
Absorbance 0.1 max. (0.1% solution) at 280nm, 0.4 max. (0.1% solution) at 230nm
Color White
CAS 151-21-3, 7647-14-5, 7757-82-6
Physical Form Solid
Melting Point 206°C
Assay Percent Range (as Fatty Alcohol Sulfate) >=99%
Linear Formula CH3(CH2)10CH2OSO3Na
pH 8.5 to 10
Quantity 500 g
Merck Index 15, 8768
Identification Pass Test
Formula Weight 288.38
Percent Purity ≥99% (≥98% C12-OH)
Grade DNase- and RNase-Free
Packaging Poly Bottle
Assay (as Fatty Alcohol Sulfate) >=99%
Chemical Name or Material Sodium Dodecyl Sulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dodecyl sulfate sodium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'