missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecyl Sulfate,Ultra Pure, 50g, MP Biomedicals
Click to view available options
Quantity:
50 g
Specifications
CAS | 151-21-3 |
Molecular Formula | C12H25NaO4S |
MDL Number | MFCD00036175 |
Quantity | 50 g |
InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
IUPAC Name | sodium;dodecyl sulfate |
Molecular Weight (g/mol) | 288.38 |
PubChem CID | 3423265 |
ChEBI | CHEBI:8984 |
Show More |
Safety and Handling
Product Identifier
- Sodium Dodecyl Sulfate, Ulta Pure
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P330-Rinse mouth.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'