missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecyl Sulfate,Ultra Pure, 100g, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| CAS | 151-21-3 |
| Quantity | 100 g |
| Molecular Formula | C12H25NaO4S |
| MDL Number | MFCD00036175 |
| InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
| SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| IUPAC Name | sodium dodecyl sulfate |
| Molecular Weight (g/mol) | 288.38 |
| PubChem CID | 3423265 |
| ChEBI | CHEBI:8984 |
| Show More |
Safety and Handling
Product Identifier
- Sodium Dodecyl Sulfate, ultra pure
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : sodium dodecyl sulphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction