missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate (SDS), ≥95%

Product Code. p-7082762 Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Cardboard Carton
Plastic bottle
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 15865168

Brand: Thermo Scientific J18220.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium Lauryl Sulfate,n to Dodecyl sulfate sodium salt
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Physical Form Crystalline Powder
Melting Point ∼206°C
Quantity 500 g
Packaging Plastic bottle
Linear Formula CH3(CH2)11OSO3Na
UN Number UN2926
Beilstein 3599286
IUPAC Name sodium dodecyl sulfate
Formula Weight 288.38
Percent Purity ≥95%
Chemical Name or Material Sodium Dodecyl Sulfate (SDS)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.