missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate, MP Biomedicals

Product Code. p-7112301 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
500 g
1 kg
Unit Size:
1kg
25g
500g
This item is not returnable. View return policy

Product Code. 11451652

Brand: MP Biomedicals 04811033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  • Used to solubilize and denature proteins for denaturing-PAGE
  • Most proteins bind SDS in a ratio of 1.4 g SDS per gram of protein
  • The charges intrinsic to the protein become insignificant compared to the overall negative charge provided by the bound SDS
  • The charge to mass ratio is essentially the same for each protein and will migrate in the gel based only on their size

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material SODIUM DODECYL SULFATE
Color White
Physical Form Powder
Melting Point 180°C to 210°C
Quantity 25 g
Molecular Formula C12H25NaO4S
Linear Formula C12H25O4SNa
MDL Number MFCD00036175
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
Solubility Information Soluble in water (200mg/mL - clear, faint yellow solution), and ethanol (0.1g/10 mL).
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
IUPAC Name sodium dodecyl sulfate
Molecular Weight (g/mol) 288.38
PubChem CID 3423265
ChEBI CHEBI:8984
Formula Weight 288.38
Percent Purity ≥99%
Grade Ultra Pure
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium Dodecyl Sulfate, ultra pure
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : sodium dodecyl sulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.