missing translation for 'onlineSavingsMsg'
Learn More

Sodium dodecatungstophosphate hydrate

Product Code. 11404643
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404643 25 g 25g
11414643 100 g 100g
11424643 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404643 Supplier Thermo Scientific Alfa Aesar Supplier No. A11575.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51312-42-6 | Na3O40PW12 | 2946.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51312-42-6
Molecular Formula Na3O40PW12
Molecular Weight (g/mol) 2946.12
MDL Number MFCD00036311
IUPAC Name Sodium dodecamolybdophosphate hydrate
SMILES *

Specifications

Melting Point 350°C
Quantity 25 g
Formula Weight 2946.12 (Anhydrous)
Chemical Name or Material Sodium dodecatungstophosphate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.