missing translation for 'onlineSavingsMsg'
Learn More

Sodium DL-Malate, MP Biomedicals

Product Code. 11402756 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402756 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11402756 Supplier MP Biomedicals Supplier No. 0521805010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 676-46-0
Molecular Formula C4H4Na2O5
Quantity 10 g
Synonym disodium malate, sodium malate, disodium dl-malate, dl-malic acid disodium salt, disodium 2-hydroxybutanedioate, natriummalat german, sodium dl-malate, butanedioic acid, hydroxy-, disodium salt, malic acid, disodium salt, sodium dl-maleate
InChI Key WPUMTJGUQUYPIV-UHFFFAOYSA-L
SMILES C(C(C(=O)[O-])O)C(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;2-hydroxybutanedioate
Molecular Weight (g/mol) 178.051
PubChem CID 8736
Formula Weight 178.05
Chemical Name or Material Sodium DL-Malate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.