missing translation for 'onlineSavingsMsg'
Learn More

Sodium DL-Lactate (60% w/w in H2O), TRC

Product Code. 30138343
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138343 10 g 10g
30132824 5 g 5g
17927724 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138343 Supplier Toronto Research Chemicals Supplier No. TRCS64400310G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-17-3
Molecular Formula C3 H5 O3 . Na
Molecular Weight (g/mol) 112.06
Synonym Sodium lactate, sodium 2-hydroxypropanoate, LAC
IUPAC Name sodium;2-hydroxypropanoate
SMILES [Na+].CC(O)C(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1
Formula Weight 112.0136
Concentration or Composition (by Analyte or Components) Lactate 60 %
Chemical Name or Material Lactate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.