missing translation for 'onlineSavingsMsg'
Learn More

Sodium DL-lactate, 60% w/w aq. soln.

Product Code. p-7051557
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 11324278

Brand: Thermo Scientific Alfa Aesar 041529.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Useful chiral synthon; building block for depsipeptides | CAS: 72-17-3 | C3H5NaO3 | 112.06 g/mol

Useful chiral synthon; building block for depsipeptides.Sodium DL-lactate, 60% w/w aq. Soln is used as a food additive, plasticizer for casein, preservative, acidity regulator, bulking agent, and in shampoo and liquid soaps. It is also used for the treatment for arrhythmias and sympathomimetics. It is useful chiral synthon and building block for depsipeptides. It is used as a corrosion inhibitor in alcohol antifreeze mixtures, flavoring agent, emulsifier, corrosion inhibitor and pH control agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-17-3
Molecular Formula C3H5NaO3
Molecular Weight (g/mol) 112.06
MDL Number MFCD00065400
InChI Key NGSFWBMYFKHRBD-UHFFFAOYSA-M
Synonym sodium lactate, lactic acid sodium salt, monosodium lactate, sodium dl-lactate, lacolin, per-glycerin, lactic acid, monosodium salt, sodium alpha-hydroxypropionate, monosodium 2-hydroxypropanoate, propanoic acid, 2-hydroxy-, monosodium salt
PubChem CID 23666456
ChEBI CHEBI:75228
IUPAC Name sodium;2-hydroxypropanoate
SMILES CC(C(=O)[O-])O.[Na+]

Specifications

Color Colorless
Density 1.326 g/mL
Odor Odorless
Linear Formula C3H5NaO3
Quantity 250 mL
Beilstein 4332999
Merck Index 14,8635
Solubility Information Miscible with water and alcohol.
Formula Weight 112.06
Concentration or Composition (by Analyte or Components) 60% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Sodium DL-lactate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.