missing translation for 'onlineSavingsMsg'
Learn More

Sodium dihydrogen phosphate monohydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number 11357027
Click to view available options
Quantity:
25 g
100 g
10049-21-5
H5NaO5P
139.00
MFCD00149208
BBMHARZCALWXSL-UHFFFAOYSA-N
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
516949
sodium;dihydrogen phosphate;hydrate
O.[Na+].OP(O)(O)=O
10049-21-5
H5NaO5P
139.00
MFCD00149208
BBMHARZCALWXSL-UHFFFAOYSA-N
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
516949
sodium;dihydrogen phosphate;hydrate
O.[Na+].OP(O)(O)=O

Sodium dihydrogen phosphate monohydrate is used as a food additive. It finds use as mordant in dyeing. It is also used in the manufacture of enamels, chemicals, pharmaceuticals, ceramics, detergents, and boiler compounds. In addition, it is used as a fireproofing agent, thickening agent, emulsifier, and as a reagent and buffer in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 100°C to H2O
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.998% (metals basis)
Solubility Information Soluble in water; insoluble in ethanol,ether and chloroform.
Formula Weight 137.99 (119.98 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Chemical Name or Material Sodium dihydrogen phosphate monohydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'