missing translation for 'onlineSavingsMsg'
Learn More

Sodium dihydrogen phosphate dihydrate, 98+%

Product Code. 11471880
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
10 kg
Unit Size:
10000g
100g
2500g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471880 500 g 500g
11461880 100 g 100g
15443037 2500 g 2500g
15433037 10 kg 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11471880 Supplier Thermo Scientific Alfa Aesar Supplier No. A11316.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13472-35-0 | H6NaO6P | 156.006 g/mol

Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates

Solubility
Soluble in water.(1 M at 20°C)

Notes
Hygroscopic. Store under inert gas. Incompatible with oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H6NaO6P
MDL Number MFCD00149209
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
Synonym sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
PubChem CID 23673460
IUPAC Name sodium;dihydrogen phosphate;dihydrate

Specifications

Quantity 500 g
Linear Formula NaH2PO4·2H2O
Merck Index 14,8660
Solubility Information Soluble in water.(1 M at 20°C)
SMILES O.O.OP(=O)(O)[O-].[Na+]
Molecular Weight (g/mol) 156.006
Formula Weight 156.01 (119.98 Anhydrous)
Percent Purity 99%
Sensitivity Hygroscopic
Density 1.91
Chemical Name or Material Sodium dihydrogen phosphate dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.