missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dihydrogen Orthophosphate Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
264.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
264.73 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
264.73 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Specifications
Melting Point | 60°C |
Color | White |
pH | 4.5 |
Physical Form | Solid |
Quantity | 5 kg |
Formula Weight | 156.01g/mol |
Packaging | Plastic keg |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'