missing translation for 'onlineSavingsMsg'
Learn More

Sodium diformamide, 98%

Product Code. 10321853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10321853 5 g Glass bottle 5g
10726383 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10321853 Supplier Thermo Scientific Acros Supplier No. 380430050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18197-26-7 | C2H2NNaO2 | 95.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18197-26-7
Molecular Formula C2H2NNaO2
Molecular Weight (g/mol) 95.03
MDL Number MFCD00145302
InChI Key QJXDSDLNUKLDBP-UHFFFAOYSA-M
Synonym sodium diformylamide, diformylamide sodium salt, diformylimide sodium salt, sodium diformylazanide, sodium diformamide, diformylamino sodium, diformylsodium, acmc-209eii, sodium dimethanoylazanide, ksc538a2h
PubChem CID 10877056
IUPAC Name sodium;diformylazanide
SMILES C(=O)[N-]C=O.[Na+]

Specifications

Color Beige
Physical Form Powder
Quantity 5 g
Assay Percent Range 98%
Linear Formula NaN(COH)2
Formula Weight 95.03
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Sodium diformamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.