missing translation for 'onlineSavingsMsg'
Learn More

Sodium Diethyldithiocarbamate Trihydrate, 99+%, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10070530
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
20624-25-3
C5H16NNaO3S2
225.297
150617
WWGXHTXOZKVJDN-UHFFFAOYSA-M
sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech
517546
sodium;N,N-diethylcarbamodithioate;trihydrate
CCN(CC)C(=S)[S-].O.O.O.[Na+]
This item is not returnable. View return policy
20624-25-3
C5H16NNaO3S2
225.297
150617
WWGXHTXOZKVJDN-UHFFFAOYSA-M
sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech
517546
sodium;N,N-diethylcarbamodithioate;trihydrate
CCN(CC)C(=S)[S-].O.O.O.[Na+]
Melting Point 95°C
Color White
pH 7
Physical Form Solid
Quantity 100 g
Formula Weight 225.31g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Diethyldithiocarbamic acid, sodium salt trihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'