missing translation for 'onlineSavingsMsg'
Learn More

Sodium diethyldithiocarbamate trihydrate, 98%

Product Code. p-7022914
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442628 100 g 100g
11452628 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11442628 Supplier Thermo Scientific Alfa Aesar Supplier No. A15898.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20624-25-3 | C5H16NNaO3S2 | 225.297 g/mol

Sodium diethyldithiocarbamate trihydrate is a spin trapping reagent used in conjunction with ferrous ion to detect nitric oxide in brain, kidney, liver and other tissues. It is used as sensitive reagent for the determination of copper, rubber vulcanization accelerator and finds application in the determination of other metals such as zinc, cobalt, platinum and palladium. Furthermore, it is an inhibitor of superoxide dismutase, ascorbate oxidase and other enzymes as well as inhibits the induction of macrophage nitric oxide synthase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium diethyldithiocarbamate trihydrate is a spin trapping reagent used in conjunction with ferrous ion to detect nitric oxide in brain, kidney, liver and other tissues. It is used as sensitive reagent for the determination of copper, rubber vulcanization accelerator and finds application in the determination of other metals such as zinc, cobalt, platinum and palladium. Furthermore, it is an inhibitor of superoxide dismutase, ascorbate oxidase and other enzymes as well as inhibits the induction of macrophage nitric oxide synthase.

Solubility
Soluble in water, ethanol, methanol and acetone. Insoluble in ether.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20624-25-3
Molecular Formula C5H16NNaO3S2
Molecular Weight (g/mol) 225.297
MDL Number MFCD00150617
InChI Key WWGXHTXOZKVJDN-UHFFFAOYSA-M
Synonym sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech
PubChem CID 517546
IUPAC Name sodium;N,N-diethylcarbamodithioate;trihydrate
SMILES CCN(CC)C(=S)[S-].O.O.O.[Na+]

Specifications

Melting Point 94°C to 102°C
Quantity 100 g
Beilstein 3920507
Sensitivity Hygroscopic
Merck Index 14,3378
Solubility Information Soluble in water,ethanol,methanol and acetone. Insoluble in ether.
Formula Weight 225.31 (171.28 Anhydrous)
Percent Purity 98%
Chemical Name or Material Sodium diethyldithiocarbamate trihydrate
compliance-icons compliance-icons
Product Identifier
  • Sodium diethyldithiocarbamate trihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Diethylcarbamodithioic acid sodium salttrihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.