missing translation for 'onlineSavingsMsg'
Learn More

Sodium dicyanamide, 97%, pure

Product Code. 10435662 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10435662 100 g Plastic bottle 100g
10510902 25 g Plastic bottle 25g
10045943 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10435662 Supplier Thermo Scientific Acros Supplier No. 176341000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1934-75-4 | C2N3Na | 89.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1934-75-4
Molecular Formula C2N3Na
Molecular Weight (g/mol) 89.03
MDL Number MFCD00003535
InChI Key IXBPPZBJIFNGJJ-UHFFFAOYSA-N
Synonym sodium dicyanamide, sodium dicyanoazanide, sodium dicyanoamide, unii-i9c96i1mre, cyanamide, cyano-, sodium salt, cyanamide,cyano-,sodium salt, i9c96i1mre, cyanamide, n-cyano-, sodium salt 1:1, dicyanamide, sodium salt, nadca
PubChem CID 4350929
SMILES [Na+].N#C[N-]C#N

Specifications

Melting Point >300°C
Color Pink or White
Infrared Spectrum Authentic
Assay Percent Range 2% max. (NaCl) (Argentometry)
Packaging Plastic bottle
Linear Formula NaN(CN)2
Quantity 100 g
Beilstein 03, III, 154
Solubility Information Solubility in water: 260g/L (30°C). Other solubilities: insoluble in aceton,etoh and ethyl acetate
IUPAC Name sodium;dicyanoazanide
Formula Weight 89.03
Percent Purity 97%
Grade Pure
Physical Form Powder
Chemical Name or Material Sodium dicyanamide, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Sodium dicyanamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.