missing translation for 'onlineSavingsMsg'
Learn More

Sodium dicyanamide, 96%

Product Code. 11440187
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440187 25 g 25g
11450187 100 g 100g
11460187 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440187 Supplier Thermo Scientific Alfa Aesar Supplier No. B21634.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 1934-75-4 | C2N3Na | 89.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1934-75-4
Molecular Formula C2N3Na
Molecular Weight (g/mol) 89.03
MDL Number MFCD00003535
InChI Key IXBPPZBJIFNGJJ-UHFFFAOYSA-N
Synonym sodium dicyanamide, sodium dicyanoazanide, sodium dicyanoamide, unii-i9c96i1mre, cyanamide, cyano-, sodium salt, cyanamide,cyano-,sodium salt, i9c96i1mre, cyanamide, n-cyano-, sodium salt 1:1, dicyanamide, sodium salt, nadca
PubChem CID 4350929
IUPAC Name sodium;dicyanoazanide
SMILES [Na+].N#C[N-]C#N

Specifikationer

Melting Point >300°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 89.03
Percent Purity 96%
Chemical Name or Material Sodium dicyanamide

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.