missing translation for 'onlineSavingsMsg'
Learn More

Sodium dicyanamide, 96%

Product Code. 11450187
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450187 100 g 100g
11440187 25 g 25g
11460187 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450187 Supplier Thermo Scientific Alfa Aesar Supplier No. B21634.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1934-75-4 | C2N3Na | 89.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1934-75-4
Molecular Formula C2N3Na
Molecular Weight (g/mol) 89.03
MDL Number MFCD00003535
InChI Key IXBPPZBJIFNGJJ-UHFFFAOYSA-N
Synonym sodium dicyanamide, sodium dicyanoazanide, sodium dicyanoamide, unii-i9c96i1mre, cyanamide, cyano-, sodium salt, cyanamide,cyano-,sodium salt, i9c96i1mre, cyanamide, n-cyano-, sodium salt 1:1, dicyanamide, sodium salt, nadca
PubChem CID 4350929
IUPAC Name sodium;dicyanoazanide
SMILES [Na+].N#C[N-]C#N

Specifications

Melting Point >300°C
Quantity 100 g
Sensitivity Hygroscopic
Formula Weight 89.03
Percent Purity 96%
Chemical Name or Material Sodium dicyanamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.