missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dichromate Dihydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Sodium Dichromate Dihydrate |
| CAS | 7789-12-0 |
| Molecular Formula | Cr2Na2O7.2(H2O) |
| Quantity | 25 g |
| Synonym | Dichromic Acid (H2Cr2O7) Disodium Salt Dihydrate, Dichromic Acid Disodium Salt Dihydrate, Disodium Dichromate Dihydrate, Sodium Chromate (Na2Cr2O7) Dihydrate, Sodium Dichromate (Na2Cr2O7) Dihydrate, Sodium Dichromate Dihydrate, Sodium Dichromate Dihydrate (Na2Cr2O7.2H2O) |
| InChI Formula | InChI=1S/2Cr.2Na.2H2O.7O/h;;;;2*1H2;;;;;;;/q;;2*+1;;;;;;;;2*-1 |
| SMILES | O.O.[Na+].[Na+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] |
| IUPAC Name | disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate |
| Molecular Weight (g/mol) | 261.97 |
| Formula Weight | 297.8461 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?