missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dichromate dihydrate, ACS, 99.5-100.5%, Thermo Scientific Chemicals
17.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
It is used as a strong oxidizing agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 100°C-2H2O |
Color | Red |
Physical Form | Crystalline |
Quantity | 100 g |
Assay Percent Range | 99.5-100.5% |
Solubility Information | Soluble in water. Insoluble in alcohol. |
Formula Weight | 298.00 (261.97 Anhydrous) |
Grade | ACS |
Sensitivity | Hygroscopic |
Appearance | Red crystalline |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'