missing translation for 'onlineSavingsMsg'
Learn More

Sodium dichromate dihydrate, 99%

Product Code. 10216610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10216610 1 kg Plastic Bottle 1kg
10348640 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10216610 Supplier Thermo Scientific Acros Supplier No. 219240010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 7789-12-0 | Cr2Na2O7·2H2O | 298 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-12-0
Molecular Formula Cr2Na2O7·2H2O
Molecular Weight (g/mol) 298
MDL Number MFCD00149166
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
Synonym sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
PubChem CID 108005
IUPAC Name disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
SMILES O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]

Specifications

Boiling Point 400.0°C
Melting Point 357.0°C
Color Orange
pH 3.5 to 3.9 (5% soln.)
Physical Form Crystals or Needles
Quantity 1 kg
Assay Percent Range 98.5% min. (Iodometry)
Linear Formula Na2Cr2O7·2H2O
Fieser 01,1059; 03,265
Merck Index 15, 8744
Solubility Information Solubility in water: 730g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 298
Percent Purity 99%
Packaging Plastic Bottle
Chemical Name or Material Sodium dichromate dihydrate, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium dichromate dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1B
  • Oxidising solid Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H360FD-May damage fertility. May damage the unborn child.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H272-May intensify fire; oxidiser.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.