missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dichloroacetate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Sodium dichloroacetate finds medicinal application in removing warts and other skin growths. It is used as an antidote for lactic acidosis and in the treatment of stroke. It is studied as potential drugs, since it inhibits the enzyme pyruvate dehydrogenase kinase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium dichloroacetate finds medicinal application in removing warts and other skin growths. It is used as an antidote for lactic acidosis and in the treatment of stroke. It is studied as potential drugs, since it inhibits the enzyme pyruvate dehydrogenase kinase.
Solubility
Soluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Sodium dichloroacetate finds medicinal application in removing warts and other skin growths. It is used as an antidote for lactic acidosis and in the treatment of stroke. It is studied as potential drugs, since it inhibits the enzyme pyruvate dehydrogenase kinase.
Solubility
Soluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 197°C to 199°C (decomposition) |
Quantity | 250 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 150.93 |
Percent Purity | 97% |
Chemical Name or Material | Sodium dichloroacetate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'