missing translation for 'onlineSavingsMsg'
Learn More

Sodium dichloroacetate, 96%

Product Code. 11434867
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434867 10 g 10g
11444867 50 g 50g
11454867 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434867 Supplier Thermo Scientific Alfa Aesar Supplier No. B21827.09
  • ONLINE EXCLUSIVE PRICE
    £45.00
    £39.65 / 10g
    Save £5.35
    11% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2156-56-1 | C2HCl2NaO2 | 150.92 g/mol

Sodium dichloroacetate finds medicinal application in removing warts and other skin growths. It is used as an antidote for lactic acidosis and in the treatment of stroke. It is studied as potential drugs, since it inhibits the enzyme pyruvate dehydrogenase kinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dichloroacetate finds medicinal application in removing warts and other skin growths. It is used as an antidote for lactic acidosis and in the treatment of stroke. It is studied as potential drugs, since it inhibits the enzyme pyruvate dehydrogenase kinase.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2156-56-1
Molecular Formula C2HCl2NaO2
Molecular Weight (g/mol) 150.92
MDL Number MFCD00070489
InChI Key LUPNKHXLFSSUGS-UHFFFAOYSA-M
Synonym sodium dichloroacetate, sodium 2,2-dichloroacetate, dichloroacetic acid sodium salt, acetic acid, dichloro-, sodium salt, ceresine, dichloroacetate sodium, dichloroctan sodny czech, sodium dichloroacetate usan, ccris 7697, dichloroacetic acid, sodium salt
PubChem CID 517326
IUPAC Name sodium;2,2-dichloroacetate
SMILES [Na+].[O-]C(=O)C(Cl)Cl

Specifications

Melting Point 197°C to 199°C (decomposition)
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 150.93
Percent Purity 97%
Chemical Name or Material Sodium dichloroacetate
compliance-icons
Product Identifier
  • Sodium dichloroacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium dichloroacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.