missing translation for 'onlineSavingsMsg'
Learn More

Sodium Diatrizoate, MP Biomedicals™

Product Code. 11416302 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416302 10 g 10g
11426302 50 g 50g
11436302 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416302 Supplier MP Biomedicals Supplier No. 0210290810

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a density gradient reagent in blood cell separation. Sodium Diatrizoate, MP Biomedicals is also utilized as a radiological contrast agent. It causes partial redistribution of tight-junction-associated membrane proteins into a cytoplasmic compartme

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 737-31-5
Molecular Formula C11H8I3N2NaO4
Molecular Weight (g/mol) 635.898
InChI Key ZEYOIOAKZLALAP-UHFFFAOYSA-M
Synonym diatrizoate sodium, sodium diatrizoate, sodium amidotrizoate, hypaque sodium, triombrin, triombrine, hypaque, vascoray, hypaque cysto, hpaque-cysto
PubChem CID 23672589
ChEBI CHEBI:53692
IUPAC Name sodium;3,5-diacetamido-2,4,6-triiodobenzoate
SMILES CC(=O)NC1=C(C(=C(C(=C1I)C(=O)[O-])I)NC(=O)C)I.[Na+]

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Soluble in water (350mg/mL).
Formula Weight 635.9
Percent Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.