missing translation for 'onlineSavingsMsg'
Learn More

Sodium deoxycholate monohydrate, 98%, Thermo Scientific Chemicals

Product Code. 11405636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405636 100 g 100g
11495626 25 g 25g
11415636 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405636 Supplier Thermo Scientific Alfa Aesar Supplier No. B20759.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Sodium deoxycholate monohydrate is used as a solubilization agent for estrogen synthetase from human placental microsomes. It is also used to study the immobilization of lipoxygenase in an alginate-silicate gel matrix and to investigate the in vitro dissolution of gall stones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium Deoxycholate Monohydrate is used as a solubilization agent for estrogen synthetase from human placental microsomes. It is also used to study the immobilization of lipoxygenase in an alginate-silicate gel matrix and to investigate the in vitro dissolution of gall stones.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-92-6
Molecular Formula C24H41NaO5
Molecular Weight (g/mol) 432.58
MDL Number MFCD00150750 MFCD00064139
InChI Key NDVHNZISGVSFMP-WTCAICSISA-M
Synonym 3alpha,5beta,12alpha-3,12-dihydroxycholan-24-oic acid monosodium salt monohydrate
PubChem CID 67096373
SMILES O.[Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])CC[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC([O-])=O

Specifications

Melting Point >300°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 0.98
Beilstein 3643164
Solubility Information Soluble in water.
IUPAC Name sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate hydrate
Formula Weight 432.58 (414.58 Anhydrous)
Chemical Name or Material Sodium Deoxycholate Monohydrate
compliance-icons
Product Identifier
  • Sodium deoxycholate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Sodium deoxycholatemonohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.