missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dehydrocholate, TRC

Product Code. 30424003
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424003 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424003 Supplier Toronto Research Chemicals Supplier No. TRCS08392310G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dehydrocholic acid sodium salt
CAS 145-41-5
Molecular Formula C24 H33 O5 . Na
Quantity 10 g
Synonym 3,7,12-Trioxo-5β-cholan-24-oic acid sodium salt, Dilabil sodium
InChI Formula InChI=1S/C24H34O5.Na/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26;/h13-14,16-18,22H,4-12H2,1-3H3,(H,28,29);/q;+1/p-1/t13-,14+,16-,17+,18+,22+,23+,24-;/m1./s1
SMILES [Na+].C[C@H](CCC(=O)[O-])[C@H]1CC[C@H]2[C@H]3[C@H](CC(=O)[C@]12C)[C@@]4(C)CCC(=O)C[C@H]4CC3=O
IUPAC Name sodium;(4R)-4-[(5S,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-3,7,12-trioxo-1,2,4,5,6,8,9,11,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]pentanoate
Molecular Weight (g/mol) 424.5
Formula Weight 424.2226
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.