missing translation for 'onlineSavingsMsg'
Learn More

Sodium D-gluconate, 97%

Product Code. 11408703
Change view
Click to view available options
Quantity:
2500 g
10 kg
Unit Size:
10000g
2500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408703 10 kg 10000g
11498693 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408703 Supplier Thermo Scientific Alfa Aesar Supplier No. A10464.0C
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 527-07-1 | C6H11NaO7 | 218.137 g/mol

Sodium D-gluconate is a good chelator at alkaline pH, with chelating power relatively better than EDTA, NTA and other chelators. It has wide range of industrial applications which include textile dyeing, water treatments, printing and as cleaning agent for glass bottles. It also finds use as derusting agent in metallurgy and as additive in cement. It is used in food stuffs as bitterness inhibitor and as electrolyte replenisher.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium D-gluconate is a good chelator at alkaline pH, with chelating power relatively better than EDTA, NTA and other chelators. It has wide range of industrial applications which include textile dyeing, water treatments, printing and as cleaning agent for glass bottles. It also finds use as derusting agent in metallurgy and as additive in cement. It is used in food stuffs as bitterness inhibitor and as electrolyte replenisher.

Solubility
Very soluble in water; sparingly soluble in alcohol; insoluble in ether.

Notes
In general it is stable, non corrosive, non-toxic, readily biodegradable and incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-07-1
Molecular Formula C6H11NaO7
Molecular Weight (g/mol) 218.137
MDL Number MFCD00064210
InChI Key UPMFZISCCZSDND-JJKGCWMISA-M
Synonym sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t
PubChem CID 23672301
ChEBI CHEBI:84997
IUPAC Name sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+]

Specifications

Melting Point ∽206°C (decomposition)
Quantity 10 kg
Beilstein 3919651
Solubility Information Very soluble in water; sparingly soluble in alcohol; insoluble in ether.
Formula Weight 218.14
Chemical Name or Material Sodium D-gluconate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.