missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyclopentadienide, 2-3M in THF

Product Code. 11336389
Change view
Click to view available options
Quantity:
0.1 mol
0.5 mol
Unit Size:
0.1mol
0.5mol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336389 0.5 mol 0.5mol
11326389 0.1 mol 0.1mol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336389 Supplier Thermo Scientific Alfa Aesar Supplier No. 088300.JT
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent for the preparation of metallocenes | CAS: 4984-82-1 | C5H5Na | 88.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4984-82-1
Molecular Formula C5H5Na
Molecular Weight (g/mol) 88.085
MDL Number MFCD00012161
InChI Key JQUZTGJSSQCTPV-UHFFFAOYSA-N
Synonym cyclopentadienylsodium, sodium cyclopenta-1,3-dien-1-ide, sodium cyclopentadienide, 2.0m solution in tetrahydrofuran
PubChem CID 21915903
IUPAC Name sodium;cyclopenta-1,3-diene
SMILES C1C=CC=[C-]1.[Na+]

Specifications

Color Pink to Red
Density 0.938 g/mL
Flash Point −17°C (1°F)
Odor Ether-like
Quantity 0.5 mol
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 88.09
Concentration or Composition (by Analyte or Components) 2 to 3M in THF
Physical Form Liquid
Chemical Name or Material Sodium cyclopentadienide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.