missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanide, Reagent Grade, 97%, Honeywell™

Product Code. p-7120784 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15645220 5 g Poly Bottle 5g
15625220 100 g Poly Bottle 100g
15635220 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15645220 Supplier Honeywell Research Chemicals Supplier No. 3809705G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
Synonym sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
PubChem CID 8929
IUPAC Name sodium;cyanide
SMILES [Na+].[C-]#N

Specifications

Boiling Point 1500°C
Melting Point 563.7°C
Color White
pH 11 to 12 (49g/L at 25°C)
Quantity 5 g
Linear Formula NaCN
UN Number UN1689
Beilstein 3587243
Solubility Information In water 49g/L at 20°C
Formula Weight 49.01g/mol
Percent Purity 97%
Packaging Poly Bottle
Appearance Crystalline powder or granules or chunks
Vapor Pressure 1mmHg (817°C)
Density Vapor Density: 1.7 (vs air)
Chemical Name or Material Sodium cyanide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.