missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanide, ACS Reagent, ≥95.0%, Honeywell Fluka™

Product Code. p-7120783 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15671750

Brand: Honeywell-Fluka 205222100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

ACS Reagent,≥95.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
Synonym sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
PubChem CID 8929
IUPAC Name sodium;cyanide
SMILES [Na+].[C-]#N

Specifications

Boiling Point 1407°C
Melting Point 563°C
Color Colorless
pH 11 to 12 (49g/L at 25°C)
Quantity 100 g
Linear Formula NaCN
UN Number UN1689
Beilstein 3587243
Solubility Information Soluble in water
Formula Weight 49.01g/mol
Percent Purity ≥95.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Crystals
Vapor Pressure 1mmHg (817°C)
Density 1.6g/cu.cm; Vapor Density: 1.7 (vs air)
Chemical Name or Material Sodium Cyanide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.