missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanide, ACS Reagent, ≥95.0%, Solstice

Product Code. 15671750 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15671750 100 g Poly Bottle 100g
15691750 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15671750 Supplier Honeywell-Fluka Supplier No. 205222100G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

ACS Reagent,≥95.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
Synonym sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
PubChem CID 8929
IUPAC Name sodium;cyanide
SMILES [Na+].[C-]#N

Specifications

Boiling Point 1407°C
Melting Point 563°C
Color Colorless
pH 11 to 12 (49g/L at 25°C)
Quantity 100 g
Linear Formula NaCN
UN Number UN1689
Beilstein 3587243
Solubility Information Soluble in water
Formula Weight 49.01g/mol
Percent Purity ≥95.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Crystals
Vapor Pressure 1mmHg (817°C)
Density 1.6g/cu.cm; Vapor Density: 1.7 (vs air)
Chemical Name or Material Sodium Cyanide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.