missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanide, ACS, 95% min, Thermo Scientific Chemicals

Catalog Number 11326449
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
143-33-9
CNNa
49.01
MFCD00003523
MNWBNISUBARLIT-UHFFFAOYSA-N
sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
8929
sodium;cyanide
[Na+].[C-]#N
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
143-33-9
CNNa
49.01
MFCD00003523
MNWBNISUBARLIT-UHFFFAOYSA-N
sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
8929
sodium;cyanide
[Na+].[C-]#N

Sodium cyanide is used as a starting material for the preparation of Reissert compounds, cyanogen bromide, cyanuric chloride and cyanogen chloride. It acts as a catalyst for the aminolysis of esters to primary amides. Furthermore, it is used for fumigation, in electroplating and for extracting gold and silver in mining industry. In organic synthesis, it is involved in the cyanation reaction of alkyl halides under phase transfer conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point ca 1500°C
Melting Point 563°C
Color Gray
Physical Form Granules
Quantity 100 g
Assay Percent Range 95% min.
Beilstein 3587243
Solubility Information Soluble in water,ammonia,methanol and ethanol. Slightly soluble in dimethylformamide. Insoluble in dimethylsulphoxide.
Formula Weight 49.01
Grade ACS
Odor Weak, characteristic
Sensitivity Hygroscopic
Appearance White powder
Density 1.61 g/mL
Chemical Name or Material Sodium cyanide, For analysis ACS
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'