missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanate, 95%

Product Code. 11419010
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419010 500 g 500g
11409010 100 g 100g
11499000 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419010 Supplier Thermo Scientific Alfa Aesar Supplier No. A12413.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 917-61-3 | CNNaO | 65.007 g/mol

Sodium cyanate is an ideal nucleophile, and these nucleophilic properties make it a major contributor to the stereospecificity in certain reactions such as in the production of chiral oxazolidone. Sodium cyanate is a useful reagent in producing asymmetrical urea that have a range of biological activity mostly in aryl isocyanate intermediates. Such intermediates as well as sodium cyanate have been used in medicine as a means of counterbalancing carcinogenic effects on the body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium cyanate is an ideal nucleophile, and these nucleophilic properties make it a major contributor to the stereospecificity in certain reactions such as in the production of chiral oxazolidone. Sodium cyanate is a useful reagent in producing asymmetrical urea that have a range of biological activity mostly in aryl isocyanate intermediates. Such intermediates as well as sodium cyanate have been used in medicine as a means of counterbalancing carcinogenic effects on the body.

Solubility
Soluble in water 11.6 g/100 mL (25°C). Slightly soluble in ammonia, benzene. Insoluble in diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917-61-3
Molecular Formula CNNaO
Molecular Weight (g/mol) 65.007
MDL Number MFCD00011121
InChI Key ZVCDLGYNFYZZOK-UHFFFAOYSA-M
Synonym sodium cyanate, zassol, cyanic acid, sodium salt, sodiumcyanate, cyansan, weecon, sodium isocyanate, san-cyan, unii-8ufs3jrv8p, naocn
PubChem CID 517096
ChEBI CHEBI:38906
IUPAC Name sodium;cyanate
SMILES C(#N)[O-].[Na+]

Specifications

Melting Point 550°C
Density 1.89
Quantity 500 g
Beilstein 3594963
Merck Index 14,8604
Solubility Information Soluble in water 11.6g/100 mL (25°C). Slightly soluble in ammonia,benzene. Insoluble in diethyl ether.
Formula Weight 65.01
Percent Purity 95%
Chemical Name or Material Sodium cyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.