missing translation for 'onlineSavingsMsg'
Learn More

Sodium copper chlorophyllin B, MedChemExpress

Product Code. 16472389
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472389 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16472389 Supplier Medchem Express Supplier No. HYB2226
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sodium copper chlorophyllin B exerts antiviral activities against Influenza virus and HIV with IC50s of 50 to 100 μM for both of them.

Sodium copper chlorophyllin B exerts antiviral activities against Influenza virus and HIV with IC50s of 50 to 100 μM for both of them.

Specifications

Chemical Name or Material Sodium copper chlorophyllin B
Physical Form Solid
CAS 28302-36-5
Color Black
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C34H29CuN4Na3O7
For Use With (Application) COVID-19-anti-virus
Solubility Information H2O : 10 mg/mL (13.55 mM; Need ultrasonic) ∣DMSO : 6.67 mg/mL (9.04 mM; Need ultrasonic)
SMILES O=C(O[Na])CC1=C(C(C(O[Na])=O)=C(C)C2=CC3=[N]4C(C(C=O)=C3CC)=CC5=C(C=C)C(C)=C6[N-]75)[N-]2[Cu+2]47[N](C8=C6)=C1C(C8C)CCC(O[Na])=O
Molecular Weight (g/mol) 738.13
Formula Weight 738.13
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.