missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate Dihydrate, TRC

Product Code. 30413941
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30413941 100 g -
30410726 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30413941 Supplier Toronto Research Chemicals Supplier No. TRCS615210100G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6132-04-3
Molecular Formula C6H5O7.3Na.2H2O
Molecular Weight (g/mol) 294.1
Synonym Sodium citrate, Trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, 1,2,3-Propanetricarboxylic acid, 2-hydroxy-, sodium salt, hydrate (1:3:2), 1,2,3-Propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate (9CI), Citric acid, trisodium salt, dihydrate (8CI), N 1560, Sodium citrate dihydrate, Trisodium citrate dihydrate
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES O.O.[Na+].[Na+].[Na+].OC(CC(=O)[O-])(CC(=O)[O-])C(=O)[O-]

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3
Formula Weight 293.994
Percent Purity >95%
Chemical Name or Material Sodium Citrate Dihydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.