missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Citrate Dihydrate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Sodium Citrate Dihydrate
Specifications
Specifications
| Chemical Name or Material | Sodium Citrate Dihydrate |
| CAS | 4-3-6132 |
| Analyte or Component Names | Sodium Citrate Dihydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H5 O7 . 3 Na . 2 H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Sodium citrate, Trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Sodium Citrate Dihydrate |
| InChI Formula | InChI=1 S/C6H8O7.3 Na0.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3 |
| SMILES | O.O.[Na+].[Na+].[Na+].OC(CC(=O)[O-])(CC(=O)[O-])C(=O)[O-] |
| IUPAC Name | trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction