missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate Dihydrate, Analytical Grade, Penta Chemicals

Product Code. 30020452
Change view
Click to view available options
Quantity:
500 g
1000 g
Unit Size:
1kg
500g
Product Code. Quantity unitSize
30020452 1000 g 1kg
30020657 500 g 500g
2 options
This item is not returnable. View return policy

Product Code. 30020452

Brand: PENTA CHEMICALS 1147031000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Top quality chemicals for laboratories, schools, hospitals, research institutes, and large manufacturing plants.

About us

The company PENTA was established in 1990 under the trade name Ing. Petr Švec - PENTA s.r.o.. From the very beginning, we have been a solely Czech company which is managed by the Švec family, including company founder Petr Švec.

We specialize in the manufacture and distribution of pure, laboratory, pharmaceutical and specialty chemicals, which we market under the trade name Penta. We manufacture and supply a wide range of organic and inorganic chemicals, in large volumes, with a quality ranging from technical, to pure, to A.G. and ACS.

High quality and flexibility

Thanks to the high quality of our products, and our ability to fulfil specific customer requirements, we rank among the leading suppliers of qualified chemicals in the domestic market, and more recently we have also expanded into the global market.

We manufacture the chemicals in our own modern complex, which fulfils the strictest quality and safety criteria. Our large warehouse facilities, including two automated warehouses, enable us to accomodate client requirements. We package the products in accordance with all legislative requirements, and can deliver them to the customer using our own ADR transport on request.

Environmental responsibility

Our priority is to supply the maximum quality of chemicals, reliability, and environmental awareness. Our efforts are evidenced by many awards and certificates, including ISO 9001, ISO 14001 and the Responsible Care certificate for responsible business in the chemical industry.

Chemical Identifiers

CAS 4-3-6132
Molecular Weight (g/mol) 294.1

Specifications

Physical Form Crystalline Powder, Needles or Crystals
Quantity 1000 g
Percent Purity ≥99%
Grade Analytical
Packaging Plastic Jar
Chemical Name or Material Sodium Citrate Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.