missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium citrate, 0.5M buffer soln., pH 6.5

Product Code. 15456049
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
This item is not returnable. View return policy

Product Code. 15456049

Brand: Thermo Scientific Chemicals J63888.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium citrate is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium citrate is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts. 

Solubility
Soluble in water.

Notes
20X SSC Solution (RNase-free) should be stored at room temperature. This reagent should remain stable for at least 1 year in its unopened container.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium citrate buffer soln.
Color Colorless
Concentration 0.5 M
pH 6.5
Physical Form Liquid
Quantity 500 mL
Odor Odorless
MDL Number MFCD00012462
Solubility Information Soluble in water.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.