missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium citrate, 0.5M buffer soln., pH 5.0

Product Code. 15495209
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
This item is not returnable. View return policy

Product Code. 15495209

Brand: Thermo Scientific Chemicals J62918.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Citrate, is used as an anticoagulant and also used as a biological buffer. It is often used to prepare sodium citrate buffer for antigen retrieval of tissue samples. The citrate solution is designed to break protein cross-links, thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies. Citrate has anticoagulant activity and as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citrate, is used as an anticoagulant and also used as a biological buffer. It is often used to prepare sodium citrate buffer for antigen retrieval of tissue samples. The citrate solution is designed to break protein cross-links, thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies. Citrate has anticoagulant activity and as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot.

Solubility
It is soluble in water (29.4 g/L) at 20°C. Insoluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium citrate buffer soln.
Color Colorless
Concentration 0.5 M
pH 5.0
Physical Form Liquid
Quantity 250 mL
Odor Odorless
MDL Number MFCD00012462
Solubility Information It is soluble in water (29.4g/L) at 20°C. Insoluble in ethanol.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.