missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chromate, TRC

Product Code. 30419414
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419414 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30419414 Supplier Toronto Research Chemicals Supplier No. TRCS615260100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Chromate
CAS 7775-11-3
Molecular Formula CrNa2O4
Quantity 100 g
Synonym Chromic Acid (H2CrO4) Sodium Salt, Chromium Disodium Oxide, Chromium Sodium Oxide (CrNa2O4), Disodium Chromate, Disodium Chromate (Na2CrO4), Sodium Chromate, Sodium Chromate (Na2(CrO4)), Sodium Chromate (VI), Sodium Chromium Oxide (Na2CrO4)
InChI Formula InChI=1S/Cr.2Na.4O/q;2*+1;;;2*-1
SMILES [Na+].[Na+].[O-][Cr](=O)(=O)[O-]
IUPAC Name disodium;dioxido(dioxo)chromium
Molecular Weight (g/mol) 161.97
Formula Weight 161.8997
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.