missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chromate, TRC
Specifications
Specifications
| Chemical Name or Material | Sodium Chromate |
| CAS | 7775-11-3 |
| Molecular Formula | CrNa2O4 |
| Quantity | 10 g |
| Synonym | Chromic Acid (H2CrO4) Sodium Salt, Chromium Disodium Oxide, Chromium Sodium Oxide (CrNa2O4), Disodium Chromate, Disodium Chromate (Na2CrO4), Sodium Chromate, Sodium Chromate (Na2(CrO4)), Sodium Chromate (VI), Sodium Chromium Oxide (Na2CrO4) |
| InChI Formula | InChI=1S/Cr.2Na.4O/q;2*+1;;;2*-1 |
| SMILES | [Na+].[Na+].[O-][Cr](=O)(=O)[O-] |
| IUPAC Name | disodium;dioxido(dioxo)chromium |
| Molecular Weight (g/mol) | 161.97 |
| Formula Weight | 161.8997 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?