missing translation for 'onlineSavingsMsg'
Learn More

Sodium chromate tetrahydrate, Reagent Grade

Product Code. 11386808
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386808 1 kg 1kg
11396808 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11386808 Supplier Thermo Scientific Alfa Aesar Supplier No. 013453.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

For Use As Corrosion Inhibitor, in Pigment Manufacture | CAS: 10034-82-9 | CrH8Na2O8 | 234.032 g/mol

Corrosion inhibitor, pigment manufactureSodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-82-9
Molecular Formula CrH8Na2O8
Molecular Weight (g/mol) 234.032
MDL Number MFCD00149167
InChI Key AVHUVTPOXZEIRG-UHFFFAOYSA-N
Synonym sodium chromate tetrahydrate, cro4.2na.4h2o, acmc-20ajol, disodium chromate tetrahydrate, ksc492q3d, disodium chromate 2-tetrahydrate, sodium chromate tetrahydrate, reagent grade, sodium chromate tetrahydrate, p.a
PubChem CID 22146515
IUPAC Name disodium;dioxido(dioxo)chromium;tetrahydrate
SMILES O.O.O.O.[O-][Cr](=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Yellow
Physical Form Crystals
Quantity 1 kg
Solubility Information Soluble in water.
Formula Weight 234.04 (161.97 Anhydrous)
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Appearance Yellow crystalline
Density 2.73
Chemical Name or Material Sodium chromate tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.