missing translation for 'onlineSavingsMsg'
Learn More

Sodium chromate, anhydrous, 98%, Thermo Scientific™

£18.48 - £55.55

Chemical Identifiers

CAS 7775-11-3
Molecular Formula ClNaO3
Molecular Weight (g/mol) 106.44
MDL Number MFCD00003484
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
PubChem CID 516902
ChEBI CHEBI:65242
SMILES [Na+].[O-][Cl](=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11498823
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10547.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11408833
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10547.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Sodium chromate anhydrous is used as a corrosion inhibitor in the petroleum industry, wood preservative and dyeing auxiliary in the textile industry. It is also used to prepare pigments for paints and inks. It also finds use in pharmaceuticals to determine red blood cell volume. In synthetic chemistry, it is utilized as an oxidant to convert primary alcohols and secondary alcohols to carboxylic acids and ketones respectively.

Solubility
Soluble in water. Slightly soluble in methanol and ethanol.

Notes
Hygroscopic. Incompatible with combustible materials and reducing agents.
Specifications

Chemical Identifiers

7775-11-3
106.44
YZHUMGUJCQRKBT-UHFFFAOYSA-M
CHEBI:65242
ClNaO3
MFCD00003484
516902
[Na+].[O-][Cl](=O)=O

Specifications

792°C
ClNaO3
UN3288
sodium chromate
YZHUMGUJCQRKBT-UHFFFAOYSA-M
disodium dioxochromiumbis(olate)
516902
161.97
Odorless
2.72
7775-11-3
MFCD00003484
14,8601
Soluble in water. Slightly soluble in methanol and ethanol.
[Na+].[O-][Cl](=O)=O
106.44
CHEBI:65242
98%
Hygroscopic
Sodium chromate, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H330-H334-H340-H350-H360-H372-H314-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H301-H312-H314-H317-H330-H334-H335-H340-H350-H360FD-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-889-5

missing translation for 'rtecsNumber' : GB2955000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications