missing translation for 'onlineSavingsMsg'
Learn More

Sodium chromate, 98+%, anhydrous

Product Code. 10591134 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10591134 500 g Plastic bottle 500g
10278613 5 g Plastic bottle 5g
10774184 25 g Plastic bottle 25g
10784184 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10591134 Supplier Thermo Scientific Acros Supplier No. 377875000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Sodium chromate, 98+%, CrNa2O4, CAS Number-7775-11-3, asex, atlacide, chlorate de sodium, chloric acid, sodium salt, chlorsaure, desolet, kusatol, rasikal, sodium chlorate, tumbleleaf, 500g, 98% min. (Iodometry), CHEBI:65242, Yellow, 231-889-5, 161.97, GHS Signal Word: Danger | CAS: 7775-11-3 | CrNa2O4 | 191.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-11-3
Molecular Formula CrNa2O4
Molecular Weight (g/mol) 191.97
MDL Number MFCD00003484
InChI Key PXLIDIMHPNPGMH-UHFFFAOYSA-N
Synonym sodium chromate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
PubChem CID 516902
ChEBI CHEBI:65242
SMILES [Na+].[Na+].[O-][Cr]([O-])(=O)=O

Specifications

Melting Point 792.0°C
Color Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Assay Percent Range 98% min. (Iodometry)
Linear Formula Na2CrO4
Merck Index 15, 8736
Solubility Information Solubility in water: 530g/L (20°C). Other solubilities: soluble in methanol,slightly soluble in ethanol
IUPAC Name sodium chromate
Formula Weight 161.97
Percent Purity ≥98%
Packaging Plastic bottle
Chemical Name or Material Sodium chromate, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.