missing translation for 'onlineSavingsMsg'
Learn More

Sodium chromate, 98+%, anhydrous

Product Code. p-3756711 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10278613

Brand: Thermo Scientific Acros 377870050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Sodium chromate, 98+%, CrNa2O4, CAS Number-7775-11-3, asex, atlacide, chlorate de sodium, chloric acid, sodium salt, chlorsaure, desolet, kusatol, rasikal, sodium chlorate, tumbleleaf, 500g, 98% min. (Iodometry), CHEBI:65242, Yellow, 231-889-5, 161.97, GHS Signal Word: Danger | CAS: 7775-11-3 | CrNa2O4 | 191.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-11-3
Molecular Formula CrNa2O4
Molecular Weight (g/mol) 191.97
MDL Number MFCD00003484
InChI Key PXLIDIMHPNPGMH-UHFFFAOYSA-N
Synonym sodium chromate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
PubChem CID 516902
ChEBI CHEBI:65242
SMILES [Na+].[Na+].[O-][Cr]([O-])(=O)=O

Specifications

Melting Point 792.0°C
Color Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 5 g
Assay Percent Range 98% min. (Iodometry)
Linear Formula Na2CrO4
Merck Index 15, 8736
Solubility Information Solubility in water: 530g/L (20°C). Other solubilities: soluble in methanol, slightly soluble in ethanol
IUPAC Name sodium;chlorate
Formula Weight 161.97
Percent Purity ≥98%
Packaging Plastic bottle
Chemical Name or Material Sodium chromate, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.