missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chondroitin Sulfate A, TRC

Product Code. 17963975
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
Unit Size:
100g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963975 1 g 1g
17962415 100 g 100g
17951335 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17963975 Supplier Toronto Research Chemicals Supplier No. TRCS6993431G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39455-18-0
Molecular Formula (C14 H19 N O14 S . x Na)n
Molecular Weight (g/mol) 463.37
Synonym Sodium chondroitin 4-sulfate
SMILES [Na+].[*]O[C@H]1[C@H](O)[C@@H](O)[C@H](O[C@@H]2[C@@H](NC(=O)C)[C@H]([*])O[C@H](CO)[C@@H]2OS(=O)(=O)[O-])O[C@@H]1C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H21NO15S/c1-2(15)14-3-8(7(19)13(28-11(3)22)29-30(23,24)25)26-12-6(18)4(16)5(17)9(27-12)10(20)21/h3-9,11-13,16-19,22H,1H3,(H,14,15)(H,20,21)(H,23,24,25)/t3-,4+,5+,6-,7-,8-,9+,11-,12-,13-/m1/s1
Formula Weight 499.0608
Chemical Name or Material Chondroitin sulfate A sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.