missing translation for 'onlineSavingsMsg'
Learn More

Sodium cholate hydrate, 99%, Thermo Scientific Chemicals

Product Code. p-7023480
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11458936

Brand: Thermo Scientific Alfa Aesar A17074.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Biochemical solubilizing agent Sodium cholate hydrate is a non-denaturing detergent used for the extraction of membrane proteins. It is involved in the study to assess infiltration of pressurized promoter solutions in a mesoporous silica. Further, it serves as a biochemical solubilizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biochemical solubilizing agent Sodium cholate hydrate is a non-denaturing detergent used for the extraction of membrane proteins. It is involved in the study to assess infiltration of pressurized promoter solutions in a mesoporous silica. Further, it serves as a biochemical solubilizing agent.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206986-87-0
Molecular Formula C24H39NaO5
Molecular Weight (g/mol) 430.56
MDL Number MFCD00064138 MFCD00003672
InChI Key NRHMKIHPTBHXPF-TUJRSCDTSA-M
PubChem CID 23679061
IUPAC Name sodium;(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate;hydrate
SMILES [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Quantity 10 g
Beilstein 3582354
Synonym sodium cholate hydrate, cholalic acid sodium salt, c24h39o5.na.h2o, sodium cholate hydrate, bioxtra, sodium cholate hydrate dried material, nt, sodium cholate hydrate, from ox and/or sheep bile, 3, a,7, a,12, a-trihydroxy-5, a-cholan-24-oic acid sodium salt, sodium cholate hydrate, suitable for cell culture, bioreagent, 4r-4-1s,2s,7s,11s,16s,5r,9r,10r,14r,15r-5,9,16-trihydroxy-2,15-dimethylte tracyclo 8.7.0.0<2,7>.0<11,15*gt; heptadec-14-yl pentanoic acid, sodium salt, hyd rate, sodium cholate hydrate, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
Solubility Information Soluble in water.
Optical Rotation +32° (c=0.5 in Water)
Formula Weight 430.56 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Sodium cholate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.