missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloroacetate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10,000 g
250 g
2500 g
500 g
Description
Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.
Solubility
Soluble in water, ether, chloroform, benzene and alcohol.
Notes
Incompatible with strong oxidizing agents.
Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.
Solubility
Soluble in water, ether, chloroform, benzene and alcohol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | ∽199°C (decomposition) |
Flash Point | 270°C (518°F) |
Assay Percent Range | 98% |
Linear Formula | CH2CO2Na |
Quantity | 500 g |
UN Number | UN2659 |
Beilstein | 3597157 |
Merck Index | 14,2112 |
Solubility Information | Soluble in water,ether,chloroform,benzene and alcohol. |
Formula Weight | 116.48 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'