missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chlorite (Technical Grade), TRC

Product Code. 30012661
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012661 50 g 50g
30004195 100 g 100g
30010769 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012661 Supplier Toronto Research Chemicals Supplier No. TRCS61519050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-19-2
Molecular Formula Cl O2 . Na
Molecular Weight (g/mol) 90.4416
Synonym Sodium Chlorite (Technical Grade), Chlorous acid, sodium salt (1:1), Chlorous acid, sodium salt (8CI,9CI), Alcide LD, Chloro-Clean, Chloro-Clean (chlorite), Dioxychlor, Neo Silox D, Silbrite 25, Silbrite 80, Sodium chlorite, Textone
IUPAC Name sodium;chlorite
SMILES [Na+].[O-]Cl=O

Specifications

Quantity 50 g
InChI Formula InChI=1S/ClHO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1
Formula Weight 89.9485
Chemical Name or Material Sodium Chlorite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.